Learn More
Cayman Chemical SarafloxacInd8hydroCLOrId 10mg
SDP
Catalog No. 502949544
Encompass_Preferred
An internal standard for the quantification of sarafloxacin by GC- or LC-MS
Catalog No. 50-294-9544 Supplier Cayman Chemical Supplier No. 3422610 MG
An internal standard for the quantification of sarafloxacin by GC- or LC-MS