Learn More
Cayman Chemical SarafloxacInhydrochLRIde 100mg
SDP
Catalog No. 502984652
Encompass_Preferred
A fluoroquinolone antibiotic; active against various clinical isolates of bacteria, including Bacteroides, Fusobacterium, Eubacterium, Actinomyces, and Peptococcus (MICs = 0.5-2 UG/ml); inhibits P. aeruginosa DNA gyrase; decreases air sac lesion severity in a chick model of E. coli-induced colisepticemia at 8 mg/kg in the drinking water 24 hours per day for five days
Catalog No. 50-298-4652 Supplier Cayman Chemical Supplier No. 20299100 MG
A fluoroquinolone antibiotic; active against various clinical isolates of bacteria, including Bacteroides, Fusobacterium, Eubacterium, Actinomyces, and Peptococcus (MICs = 0.5-2 UG/ml); inhibits P. aeruginosa DNA gyrase; decreases air sac lesion severity in a chick model of E. coli-induced colisepticemia at 8 mg/kg in the drinking water 24 hours per day for five days