Learn More
Medchemexpress LLC Saralasin TFA | 34273-10-4 | 99.18% | 1026.07 | 25 MG
SDP
Catalog No. 5000426471
Encompass_Preferred
Saralasin TFA, an octapeptide analog of angiotensin II, functions as a competitive angiotensin II receptor antagonist and exhibits partial agonist activity. It is utilized in the research of renovascular hypertension and renin-dependent (angiotensinogenic) hypertension.
  • Competitive angiotensin II receptor antagonist with a Ki value of 0.32 nM.
  • Exhibits partial agonist activity.
  • Inhibits cell growth in 3T3 and SV3T3 cells.
  • Restores potassium currents in myocytes.
  • Inhibits binding of FITC-Ang II to rat liver membrane preparation.
  • Reduces ovulation rate and prostaglandin levels in vitro.
  • Ameliorates oxidative stress and tissue injury in cerulein-induced pancreatitis in rats.
  • Increases serum renin activity in normal, conscious rats.
Catalog No. 50-004-26471 Supplier Medchemexpress LLC Supplier No. 25MGHYP0205B
Saralasin TFA, an octapeptide analog of angiotensin II, functions as a competitive angiotensin II receptor antagonist and exhibits partial agonist activity. It is utilized in the research of renovascular hypertension and renin-dependent (angiotensinogenic) hypertension.
  • Competitive angiotensin II receptor antagonist with a Ki value of 0.32 nM.
  • Exhibits partial agonist activity.
  • Inhibits cell growth in 3T3 and SV3T3 cells.
  • Restores potassium currents in myocytes.
  • Inhibits binding of FITC-Ang II to rat liver membrane preparation.
  • Reduces ovulation rate and prostaglandin levels in vitro.
  • Ameliorates oxidative stress and tissue injury in cerulein-induced pancreatitis in rats.
  • Increases serum renin activity in normal, conscious rats.