Learn More
Cayman Chemical Saralasin (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502950135
Encompass_Preferred
A peptide AT2 receptor agonist; binds to AT2 receptors (Ki = 0.38 nM); induces neurite outgrowth in NG108-15 cells at 100 nM; enhances acquisition of the conditioned avoidance response in rats at 5 µg/animal
Catalog No. 50-295-0135 Supplier Cayman Chemical Supplier No. 3287650 MG
A peptide AT2 receptor agonist; binds to AT2 receptors (Ki = 0.38 nM); induces neurite outgrowth in NG108-15 cells at 100 nM; enhances acquisition of the conditioned avoidance response in rats at 5 µg/animal