Learn More
AdipoGen Sarcophine
SDP
Supplier:  AdipoGen BVT0305M002
Encompass_Preferred
Chemical. CAS 55038-27-2. Formula C20H28O3. MW 316.4. Isolated from Sarcophyton glaucum. Acetylcholine esterase and ATPase inhibitor. Ichthyotoxin. Tumorigenesis and JB6 cell transformation inhibitor.
Catalog No. 50-256-2002