Learn More
Medchemexpress LLC HY-101037 100mg Medchemexpress, Sarcosine CAS:107-97-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101037100MG
Encompass_Preferred
Medchemexpress, HY-101037 100mg Sarcosine CAS:107-97-1 Sarcosine (N-Methylglycine; Sarcosin), an endogenous amino acid, is a competitive glycine transporter type I (GlyT1) inhibitor an N-methyl-D-aspartate (NMDA) receptor co-agonist. Sarcosine increases the glycine concentration, resulting in an indirect potentiation of the NMDA receptor. Sarcosine is commonly used for the research of schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0614