Learn More
Medchemexpress LLC Sarcosine-d3 | 118685-91-9 | MFCD01075434 | 99.1% | 92.11 g/mol | C3H4D3NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101037S150MG
Encompass_Preferred
Sarcosine-d3 is the deuterium-labeled analogue of sarcosine (N-methylglycine) used as a stable-isotope internal standard for mass spectrometry and quantitative analytical methods. Supplied in milligram quantities for laboratory research, it provides high isotopic enrichment and chemical purity for method development, calibration, and tracer studies.
  • Deuterated internal standard for mass spectrometry quantitation.
  • High isotopic enrichment (methyl-d3).
  • High chemical purity (99.05%).
  • Available in multiple milligram sizes, including 50 mg.
  • Molecular formula C3H4D3NO2; molecular weight 92.11 g/mol.
  • Supplied with COA and SDS for quality and safety verification.
Catalog No. 50-002-12560