Learn More
Medchemexpress LLC Sarcosine-d3 (N-methylglycine-d3) | 118685-91-9 | 99.1% | 92.11 g·mol^-1 | C3H4D3NO2 | 5 MG
SDP
Catalog No. 5000213233
Encompass_Preferred
Sarcosine-d3 (N-methylglycine-d3) is a deuterium-labeled analog of sarcosine provided as a high-purity analytical standard for use in mass spectrometry, tracer experiments, and quantitative studies. Supplied in small vials, the material is intended for analytical workflows that require isotopically labeled internal standards.
  • Stable isotope-labeled internal standard for mass spectrometry quantification.
  • High purity for reliable and reproducible analytical results.
  • Available in small sizes suitable for laboratory-scale use.
  • Suitable for tracer, metabolic, and quantitative studies.
  • Provided as a dry material; consult the safety data sheet for storage and handling.
Catalog No. 50-002-13233 Supplier Medchemexpress LLC Supplier No. HY101037S15MG
Sarcosine-d3 (N-methylglycine-d3) is a deuterium-labeled analog of sarcosine provided as a high-purity analytical standard for use in mass spectrometry, tracer experiments, and quantitative studies. Supplied in small vials, the material is intended for analytical workflows that require isotopically labeled internal standards.
  • Stable isotope-labeled internal standard for mass spectrometry quantification.
  • High purity for reliable and reproducible analytical results.
  • Available in small sizes suitable for laboratory-scale use.
  • Suitable for tracer, metabolic, and quantitative studies.
  • Provided as a dry material; consult the safety data sheet for storage and handling.