Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarcosine tert-butyl ester hydrochloride, 97%

Catalog No. AAH6317606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6317606 5 g
AAH6317614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6317606 Supplier Thermo Scientific Chemicals Supplier No. H6317606
Add to Cart

CAS: 5616-81-9 | C7H16ClNO2 | 181.66 g/mol

Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (0.3 gram in 2 mL MeOH).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5616-81-9
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00080936
InChI Key RNLQHMIDSCYLAK-UHFFFAOYSA-N
Synonym h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl
PubChem CID 11252350
IUPAC Name tert-butyl 2-(methylamino)acetate;hydrochloride
SMILES CC(C)(C)OC(=O)CNC.Cl

Specifications

Melting Point 137°C to 141°C
Color White
Linear Formula CH3NHCH2CO2C(CH3)3·HCl
Quantity 5 g
Solubility Information Soluble in methanol (0.3 gram in 2 mL MeOH).
Formula Weight 181.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Sarcosine tert-butyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.