Learn More
TARGETMOL CHEMICALS INC SARDOMOZIDE DIHYDROCHLOR 5MG
SDP
Catalog No. 502576735
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Sardomozide dihydrochloride (CGP 48664 dihydrochloride) is an inhibitor of S-adenosylmethionine decarboxylase (SAMDC IC50 = 5 nM). purity: 99%
Catalog No. 50-257-6735 Supplier TARGETMOL CHEMICALS INC Supplier No. T46765MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Sardomozide dihydrochloride (CGP 48664 dihydrochloride) is an inhibitor of S-adenosylmethionine decarboxylase (SAMDC IC50 = 5 nM). purity: 99%