Learn More
Medchemexpress LLC Sardomozide dihydrochloride | 138794-73-7 | 303.19 | 1 G
SDP
Catalog No. 5000351998
Encompass_Preferred
Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor. It inhibits SAMDC with an IC50 of 5 nM in cell assay, reducing intracellular SAMDC activity to 10% of control after 48 h treatment with 3 μM.
  • Inhibits S-adenosylmethionine decarboxylase (SAMDC)
  • IC50 of 5 nM for SAMDC
  • Suitable for research applications
Catalog No. 50-003-51998 Supplier Medchemexpress LLC Supplier No. HY13746B1G
Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor. It inhibits SAMDC with an IC50 of 5 nM in cell assay, reducing intracellular SAMDC activity to 10% of control after 48 h treatment with 3 μM.
  • Inhibits S-adenosylmethionine decarboxylase (SAMDC)
  • IC50 of 5 nM for SAMDC
  • Suitable for research applications