Learn More
Medchemexpress LLC Sardomozide dihydrochloride | 138794-73-7 | 99.7% | 303.19 | 25 MG
Supplier: Medchemexpress LLC 25MGHY13746B
Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor with broad-spectrum antiproliferative and antitumor activity. It targets SAMDC with an IC50 of 5 nM.
- Targets S-adenosylmethionine decarboxylase (SAMDC)
- Exhibits broad-spectrum antiproliferative and antitumor activity
- Inhibits human T24 bladder carcinoma cells (IC50 0.71 μM)
- Reduces intracellular SAMDC activity to 10% of control after 48 h treatment with 3 μM