Learn More
Selleck Chemical LLC Sardomozide dihydrochloride
SDP
Catalog No. 503138139
Encompass_Preferred
Sardomozide dihydrochloride (CGP 48664A) is a potent inhibitor of S-adenosylmethionine decarboxylase (SAMDC) with an IC50 of 5 nM It is also a potent inhibitor of polyamine oxidase and exhibits with broad spectrum antiproliferative and antitumor activity
Catalog No. 50-313-8139 Supplier Selleck Chemical LLC Supplier No. E169425MG
Sardomozide dihydrochloride (CGP 48664A) is a potent inhibitor of S-adenosylmethionine decarboxylase (SAMDC) with an IC50 of 5 nM It is also a potent inhibitor of polyamine oxidase and exhibits with broad spectrum antiproliferative and antitumor activity