Learn More
Cayman Chemical Sardomozide (hydrochloride), Purity: ≥95%, CAS: 138794-73-7, 10 mg
SDP
Catalog No. 502910349
Encompass_Preferred
An inhibitor of SAMDC (IC50 = 0.005 µM for the rat liver enzyme); selective for SAMDC over DAO (IC50 = 18 µM for the rat small intestine enzyme); inhibits proliferation of T24 cells (IC50 = 0.71 µM); decreases intracellular levels of spermidine and spermine and increases intracellular putrescine levels in L1210 cells at 3 µM; inhibits HIV-1 replication in PM1 cells at 0.2 and 0.4 µM; reduces tumor growth in a SK-MEL-24 melanoma mouse xenograft model at 0.5 and 5 mg/kg
Catalog No. 50-291-0349 Supplier Cayman Chemical Supplier No. 3490110 MG
An inhibitor of SAMDC (IC50 = 0.005 µM for the rat liver enzyme); selective for SAMDC over DAO (IC50 = 18 µM for the rat small intestine enzyme); inhibits proliferation of T24 cells (IC50 = 0.71 µM); decreases intracellular levels of spermidine and spermine and increases intracellular putrescine levels in L1210 cells at 3 µM; inhibits HIV-1 replication in PM1 cells at 0.2 and 0.4 µM; reduces tumor growth in a SK-MEL-24 melanoma mouse xenograft model at 0.5 and 5 mg/kg