Learn More
Enzo Life Sciences SARM (human) monoclonal antibody (Sarmy-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804831C100
Encompass_Preferred
Clone: Sarmy-1. Isotype: IgG2b. Immunogen: Recombinant human SARM (aa 93-292). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: IP. Formulation: Liquid. In PBS containing 10% glycerol and 0.02% sodium azide.
Catalog No. 50-200-9991