Learn More
Enzo Life Sciences SARM polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADICSA509E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human SARM. UniProt ID: Q6SZW1. Species reactivity: Human, Mouse. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (2µg/ml) Western Blot (0.5-2.0µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ion exchange purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7187