Learn More
BPS Bioscience Inc SARM1 Fluorogenic Assay Kit (Hydrolase Activity) 384 reactions
SDP
Catalog No. NC3736291
Encompass
The SARM1 Fluorogenic Assay Kit (Hydrolase Activity) is designed to measure NAD+ cleavage activity in screening and profiling applications. The SARM1 assay kit comes in a convenient 96 or 384-well format, with enough recombinant human SARM1 enzyme (amino acids 28-724), its substrate N6-etheno-NAD (e-NAD), and SARM1 assay buffer for 100 or 400 enzyme reactions. In addition, the kit includes a SARM1 inhibitor (DSRM-3716) for use as a control inhibitor.
Catalog No. NC3736291 Supplier BPS Bioscience Inc Supplier No. 782172
The SARM1 Fluorogenic Assay Kit (Hydrolase Activity) is designed to measure NAD+ cleavage activity in screening and profiling applications. The SARM1 assay kit comes in a convenient 96 or 384-well format, with enough recombinant human SARM1 enzyme (amino acids 28-724), its substrate N6-etheno-NAD (e-NAD), and SARM1 assay buffer for 100 or 400 enzyme reactions. In addition, the kit includes a SARM1 inhibitor (DSRM-3716) for use as a control inhibitor.