Learn More
Biolegend Purified anti-SARM1
Supplier:  Biolegend 696602
Encompass_Preferred
Purified anti-SARM1 [W16079A]; Isotype: Rat IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. NC1400021