Learn More
TARGETMOL CHEMICALS INC SARMUSTINE 5MG
SDP
Catalog No. 502580501
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Sarmustine (SarCNU) is an alkylating agent with anticancer activity that inhibits the growth of prostate cancer cells via p53-dependent and p53-independent pathways.Sarmustine mediates the selection of P140K methylguanine-DNA-methyltransferase-transduced human CD34(+) cells in vitro. purity: 99%
Catalog No. 50-258-0501 Supplier TARGETMOL CHEMICALS INC Supplier No. T286635MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Sarmustine (SarCNU) is an alkylating agent with anticancer activity that inhibits the growth of prostate cancer cells via p53-dependent and p53-independent pathways.Sarmustine mediates the selection of P140K methylguanine-DNA-methyltransferase-transduced human CD34(+) cells in vitro. purity: 99%