Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saroglitazar magnesium | 1639792-20-3 | 98.0% | 901.42 | 1 MG
SDP

Catalog No. 5000401007
Encompass_Preferred

Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα and moderate PPARγ activity, having EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively. It is for research use only.

  • Pparα agonist (ec50: 0.65 pm in hepg2 cells)
  • Moderate pparγ activity (ec50: 3 nm in hepg2 cells)
  • Causes dose-dependent reductions in serum triglycerides, free fatty acids, and glucose in db/db mice.
  • Shown efficacy in lowering triglycerides in wistar rats and marmosets.
  • Appears to have good potential as a therapeutic agent for dyslipidemia and diabetes.
  • Appearance: solid
  • Color: off-white to light yellow

Catalog No. 50-004-01007 Supplier Medchemexpress LLC Supplier No. HY19937A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα and moderate PPARγ activity, having EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively. It is for research use only.

  • Pparα agonist (ec50: 0.65 pm in hepg2 cells)
  • Moderate pparγ activity (ec50: 3 nm in hepg2 cells)
  • Causes dose-dependent reductions in serum triglycerides, free fatty acids, and glucose in db/db mice.
  • Shown efficacy in lowering triglycerides in wistar rats and marmosets.
  • Appears to have good potential as a therapeutic agent for dyslipidemia and diabetes.
  • Appearance: solid
  • Color: off-white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.