Learn More
Medchemexpress LLC Saroglitazar magnesium | 1639792-20-3 | 98.85% | 901.42 | 25 MG
SDP
Catalog No. 5000403055
Encompass_Preferred
Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist that exhibits predominant PPARα activity and moderate PPARγ activity, with EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively.
  • Novel peroxisome proliferator-activated receptor (PPAR) agonist
  • Predominant PPARα activity (EC50 of 0.65 pM in HepG2 cells)
  • Moderate PPARγ activity (EC50 of 3 nM in HepG2 cells)
  • Causes dose-dependent reductions in serum triglycerides (TG), free fatty acids (FFA), and glucose in db/db mice
  • Shows lipid-lowering effects in preclinical models
  • Indicated low risk of PPAR-associated side effects in humans
  • Appears to have good potential as a novel therapeutic agent for the treatment of dyslipidemia and diabetes
Catalog No. 50-004-03055 Supplier Medchemexpress LLC Supplier No. 25MGHY19937A
Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist that exhibits predominant PPARα activity and moderate PPARγ activity, with EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively.
  • Novel peroxisome proliferator-activated receptor (PPAR) agonist
  • Predominant PPARα activity (EC50 of 0.65 pM in HepG2 cells)
  • Moderate PPARγ activity (EC50 of 3 nM in HepG2 cells)
  • Causes dose-dependent reductions in serum triglycerides (TG), free fatty acids (FFA), and glucose in db/db mice
  • Shows lipid-lowering effects in preclinical models
  • Indicated low risk of PPAR-associated side effects in humans
  • Appears to have good potential as a novel therapeutic agent for the treatment of dyslipidemia and diabetes