Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saroglitazar magnesium | 1639792-20-3 | MFCD29075338 | 98.0% | 901.42 | C50H56MgN2O8S2 | 10 MG
SDP

Catalog No. 5000297755
Encompass_Preferred

Saroglitazar magnesium is the magnesium salt of saroglitazar, a small-molecule peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα activity and moderate PPARγ activity. Supplied as a light-yellow solid for research use, it is used in in vitro and preclinical studies of lipid and glucose metabolism.

  • Magnesium salt form of a dual PPARα/PPARγ agonist.
  • Useful for pharmacology and mechanistic studies of metabolic pathways.
  • Reported molecular weight 901.42 and formula C50H56MgN2O8S2.
  • Supplied as a light-yellow solid suitable for laboratory handling.
  • Available in small sample sizes for research use.

Catalog No. 50-002-97755 Supplier Medchemexpress LLC Supplier No. HY19937A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saroglitazar magnesium is the magnesium salt of saroglitazar, a small-molecule peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα activity and moderate PPARγ activity. Supplied as a light-yellow solid for research use, it is used in in vitro and preclinical studies of lipid and glucose metabolism.

  • Magnesium salt form of a dual PPARα/PPARγ agonist.
  • Useful for pharmacology and mechanistic studies of metabolic pathways.
  • Reported molecular weight 901.42 and formula C50H56MgN2O8S2.
  • Supplied as a light-yellow solid suitable for laboratory handling.
  • Available in small sample sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.