Learn More
Medchemexpress LLC HY-16730 5mg Medchemexpress, Sarolaner CAS:1398609-39-6 Purity:>98%
SDP
Catalog No. 501932777
Encompass_Preferred
Medchemexpress, HY-16730 5mg Sarolaner CAS:1398609-39-6 Sarolaner (PF-6450567) is an orally active, broad-spectrum ectoparasiticide, has efficacy against fleas and ticks on dogs, with LC 80 of 0.3 μg/mL against C. felis and an LC 100 of 0.003 μg/mL against O. turicata [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2777 Supplier Medchemexpress LLC Supplier No. HY167305MG
Medchemexpress, HY-16730 5mg Sarolaner CAS:1398609-39-6 Sarolaner (PF-6450567) is an orally active, broad-spectrum ectoparasiticide, has efficacy against fleas and ticks on dogs, with LC 80 of 0.3 μg/mL against C. felis and an LC 100 of 0.003 μg/mL against O. turicata [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.