Learn More
Sklar Instruments Sarot Forceps, Sklar 12 in., Straight, Serrated, Stainless Steel. EA.
SDP
Catalog No. 501181412 Shop All Sklar Instruments Products
Encompass_Preferred
Sarot Forceps, Sklar. 12 in., Straight, Serrated, Stainless Steel. Sklar Sarot Thoracic Forcesps are a ratcheted, finger ring instrument used as a hemostatic clamp to control blood flow. Sarot forceps are available in multiple lengths; however, shorter lengths are often used in thoracic procedures and therefore refered to as thoracic forceps. Longer lengths are commonly used in bariatric procedures and referred to as simply Sarot forceps. Sarot forceps have long straight shanks as well as long jaws that are available straight or curved. The jaws have full horizontal serrations that provide a firm clamp on vessels. This product is straight with an overall length of 12 inches.
Catalog No. 50-118-1412 Supplier Sklar Instruments Supplier No. 322265
Sarot Forceps, Sklar. 12 in., Straight, Serrated, Stainless Steel. Sklar Sarot Thoracic Forcesps are a ratcheted, finger ring instrument used as a hemostatic clamp to control blood flow. Sarot forceps are available in multiple lengths; however, shorter lengths are often used in thoracic procedures and therefore refered to as thoracic forceps. Longer lengths are commonly used in bariatric procedures and referred to as simply Sarot forceps. Sarot forceps have long straight shanks as well as long jaws that are available straight or curved. The jaws have full horizontal serrations that provide a firm clamp on vessels. This product is straight with an overall length of 12 inches.