Learn More
Sklar Instruments Sarot Needle Holder, Sklar, 10-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. EA/1.
SDP
Catalog No. 501437504 Shop All Sklar Instruments Products
Encompass_Preferred
Sarot Needle Holder, Sklar, 10-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. Sklar's Sarot Needle Holder is used for approximation suturing, often in cardiovascular procedures. It is a ratcheted, finger ring instrument that is available in long, short, and pediatric lengths. The shanks curve inward slightly near the finger rings, then extend straight out to short jaws with tips that feature cross-serrations and one central groove. The cross-serrated surface and groove ensrue a non-slip hold on the needle while the ratcheted locking mechanism provides additional security of the hold. This product is long with an overall length of 10-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7504 Supplier Sklar Instruments Supplier No. 203910
Add to Cart
Add to Cart
Sarot Needle Holder, Sklar, 10-1/4 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. Sklar's Sarot Needle Holder is used for approximation suturing, often in cardiovascular procedures. It is a ratcheted, finger ring instrument that is available in long, short, and pediatric lengths. The shanks curve inward slightly near the finger rings, then extend straight out to short jaws with tips that feature cross-serrations and one central groove. The cross-serrated surface and groove ensrue a non-slip hold on the needle while the ratcheted locking mechanism provides additional security of the hold. This product is long with an overall length of 10-1/4 inches.