Learn More
Medchemexpress LLC Sarpogrelate hydrochloride | 135159-51-2 | MFCD00887582 | >98.0% | 465.97 | C24H32ClNO6 | 100 MG
SDP
Catalog No. 5000240042
Encompass_Preferred
Sarpogrelate hydrochloride is a research-grade selective antagonist of 5-HT2 receptors used in studies of vascular disease and thrombosis. It has published pKi values for 5-HT2A, 5-HT2B, and 5-HT2C receptors and is supplied as a high-purity hydrochloride salt for in vitro research.
  • Selective 5-HT2 receptor antagonist with published pKi values.
  • Suitable for research into vascular disease and thrombosis.
  • High purity (>98.0% by HPLC), appropriate for analytical and biological studies.
  • Molecular weight 465.97 g/mol.
  • Available in multiple pack sizes, including 100 mg solid quantities.
Catalog No. 50-002-40042 Supplier Medchemexpress LLC Supplier No. HY10564100MG
Sarpogrelate hydrochloride is a research-grade selective antagonist of 5-HT2 receptors used in studies of vascular disease and thrombosis. It has published pKi values for 5-HT2A, 5-HT2B, and 5-HT2C receptors and is supplied as a high-purity hydrochloride salt for in vitro research.
  • Selective 5-HT2 receptor antagonist with published pKi values.
  • Suitable for research into vascular disease and thrombosis.
  • High purity (>98.0% by HPLC), appropriate for analytical and biological studies.
  • Molecular weight 465.97 g/mol.
  • Available in multiple pack sizes, including 100 mg solid quantities.