Learn More
Medchemexpress LLC Sarpogrelate hydrochloride | 135159-51-2 | MFCD00887582 | 99.1% | 465.97 g·mol⁻¹ | C24H32ClNO6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1056450MG
Encompass_Preferred
Sarpogrelate hydrochloride is a selective 5-HT2 receptor antagonist (CAS 135159-51-2) used as a research reagent to investigate serotonergic signaling, vascular disease, and thrombosis. Supplied as a solid with reported purity of 99.09% and molecular weight 465.97 g·mol⁻¹, it is intended for biochemical and pharmacological research applications.
  • Selective antagonist of 5-HT2A, 5-HT2B, and 5-HT2C receptors.
  • High reported purity suitable for biochemical assays.
  • Available in small research pack sizes for in-lab experiments.
  • Useful for vascular and thrombosis-related pharmacology studies.
Catalog No. 50-002-41059