Learn More
Description
This peptide corresponds to 14 amino acids near the amino terminus of SARS M protein.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | SARS Coronavirus Matrix |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Gene Alias | Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Matrix Protein; SARS-CoV2; SARS-CoV-2; SARS-M |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.