Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S2 ECD-Fc Recombinant Protein
Supplier:  Sino Biological 40590V02H100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S2 ECD) (YP_009724390.1) (Ser686-Pro1213) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. NC1825890