Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1-His Recombinant Protein (HPLC-verified), Endotoxin-Free
Catalog No. 5000084152
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) spike protein S1 Subunit (YP_009724390.1) (Val16-Arg685) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-000-84152 Supplier Sino Biological Supplier No. 40591V08HUE100
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) spike protein S1 Subunit (YP_009724390.1) (Val16-Arg685) was expressed with a polyhistidine tag at the C-terminus.