Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (R683A, R685A, F817P, A892P, A899P, A942P, K986P, V987P)-His Recombinant Protein (HPLC-verified), Endotoxin-Free
Catalog No. 5000083670
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S1+S2 ECD) (YP_009724390.1) (Met1-Pro1213(R683A, R685A, F817P, A892P, A899P, A942P, K986P, V987P)) was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-000-83670 Supplier Sino Biological Supplier No. 40589V08H4UE100
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S1+S2 ECD) (YP_009724390.1) (Met1-Pro1213(R683A, R685A, F817P, A892P, A899P, A942P, K986P, V987P)) was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.