Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S2 ECD(708S-1207E)-His Recombinant Protein, Endotoxin-Free
Catalog No. 5000083654
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S2 ECD) (YP_009724390.1) (Ser708-Glu1207) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-000-83654 Supplier Sino Biological Supplier No. 40590V08H1UE1
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S2 ECD) (YP_009724390.1) (Ser708-Glu1207) was expressed with a polyhistidine tag at the C-terminus.