Learn More
Genscript Corporation SARS-CoV-2 ADE-Causing Antibod
Catalog No. 5000057843
Encompass
GenScript SARS-CoV-2 ADE-Causing Antibody (S9HC) reacts with SARS-CoV-2 Spike protein of wild type and delta variant It can obviously enhance the SARS-COV-2 binding with ACE2
Catalog No. 50-000-57843 Supplier Genscript Corporation Supplier No. A021331
GenScript SARS-CoV-2 ADE-Causing Antibody (S9HC) reacts with SARS-CoV-2 Spike protein of wild type and delta variant It can obviously enhance the SARS-COV-2 binding with ACE2