Learn More
Genscript Corporation SARS-CoV-2 ADE-Causing Antibod
Supplier:  Genscript Corporation A02133100
Encompass
GenScript SARS-CoV-2 ADE-Causing Antibody (S9HC) reacts with SARS-CoV-2 Spike protein of wild type and delta variant It can obviously enhance the SARS-COV-2 binding with ACE2
Catalog No. 50-000-57844