Learn More
IVANO BIOSCIENCE Validated SARS-CoV-2 pseudovirus, Alpha B.1.1.7 variant, 100 µL of pseudovirus enables 100 reactions in a 96-well plate
Supplier:  IVANO BIOSCIENCE PSVSC2AP
Encompass_Preferred
Recombinant 3rd-generation lentiviral vector displaying the spike protein (S) of the SARS-CoV-2, Alpha B.1.1.7 variant. It incorporates a luciferase reporter gene.Performance: 1 µL per well (96-well plate) produces a signal-to-background ratio of 10³ RLU, according to the manufacturer’s protocol. 100 µL = 100 reactions.Validation: Validated lot-to-lot for infection and neutralization.Applications: This pseudovirus can be used to investigate virus entry, receptor recognition, and cellular tropism, to perform functional studies, to screen anti-S drugs and neutralizing antibodies, or to assess vaccine efficacy. For Research Use Only.Biosafety: Must be handled under BSL-2 conditions. Replication-incompetent and non-pathogenic due to multiple molecular safety modifications.Shipping and storage: Shipped on dry ice and stored at −80 °C. Avoid freeze/thaw cycles. Each shipment includes a complete protocol and a batch-specific quality control report.
Catalog No. 50-000-84550