Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag
Supplier:  ACROBiosystems NUNC522110MG
Encompass_Preferred
Recombinant Protein;10MG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5221) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa.
Catalog No. 50-205-5265
Add to Cart
Request a Quote