Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci SARS-CoV-2 (COVID-19) ORF7a Peptide
SDP

Catalog No. 16595418
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-418 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-418 Supplier ProSci Supplier No. 9283P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

SARS-CoV-2 (COVID-19) ORF7a Peptide is used for blocking the activity of the SARS-CoV-2 (COVID-19) ORF7a Antibody.

Specifications

Gene ID (Entrez) 43740573
Species SARS-CoV-2
Accession Number P0DTC7.1
Common Name ORF7a protein
Concentration 200 μg/mL
Content And Storage Store SARS-CoV-2 (COVID-19) ORF7a Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol ORF7a
Quantity 0.05 mg
Sequence 9 amino acids near the carboxy terminus of SARS-CoV-2 (COVID-19) ORF7a protein.
Synonym ORF7a protein, Accessory protein 7 A, Protein U122, Protein X4, ORF7a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.