Learn More
ProSci SARS-CoV-2 (COVID-19) ORF9b Peptide

Description
Specifications
Specifications
| Species | SARS-CoV-2 |
| Accession Number | P0DTD2.1 |
| Concentration | 200 μg/mL |
| Content And Storage | Store SARS-CoV-2 (COVID-19) ORF9b Peptide at -20°C, stable for one year. |
| Format | Liquid |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide |
| For Use With (Application) | Blocking Assay |
| Quantity | 0.05 mg |
| Sequence | 14 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) ORF9b protein. |
| Synonym | Accessory protein 9 b, ORF-9 b, Protein 9 b |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.