Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD, Mouse IgG2a Fc Tag
Catalog No. 502055287
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (SPD-C5251) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 52.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (Cat. No. SPD-C5251) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-205-5287 Supplier ACROBiosystems Supplier No. SPDC52511MG
Recombinant Protein;1MG;SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (SPD-C5251) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 52.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (Cat. No. SPD-C5251) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.2-3 ng/mL (QC tested).