Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein (N501Y), His Tag
Supplier:  ACROBiosystems S1NC52HG1MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S1 protein (N501Y), His Tag (S1N-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (N501Y)). N501Y mutaion was identified in both the SARS-CoV-2 variant (known as 20B/501Y.V1, VOC 202012/01, or B.1.1.7 lineage) which emerged in the United Kindom and SARS-CoV-2 variant (known as 20C/501Y.V2 or B.1.351 lineage) which emerged in South Africa.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.9 kDa.
Catalog No. 50-205-4740