Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S2 protein, His Tag
Catalog No. 502055257
Encompass_Preferred
Recombinant Protein;10MG;SARS-CoV-2 S2 protein, His Tag (S2N-C52H2) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S2 protein, His Tag (Cat. No. S2N-C52H2) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 Spike S2 Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-205-5257 Supplier ACROBiosystems Supplier No. S2NC52H210MG
Add to Cart
Add to Cart
Request a Quote
Recombinant Protein;10MG;SARS-CoV-2 S2 protein, His Tag (S2N-C52H2) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S2 protein, His Tag (Cat. No. S2N-C52H2) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 Spike S2 Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).