Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci SARS-CoV-2 (COVID-19) Spike G142D Δ143-145VYY Peptide (Omicron)
SDP

Catalog No. 16595514
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-514 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-514 Supplier ProSci Supplier No. 9793P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

SARS-CoV-2 (COVID-19) Spike G142D Δ143-145VYY Peptide (Omicron)is used for blocking the activity of theSARS-CoV-2 (COVID-19) Spike G142D Δ143-145VYY Antibody (Omicron)

SARS-CoV-2 Omicron variant, a variant of concern (VOC), known as B.1.1.529, was detected in South Africa at the end of November in 2021. However, it rapidly spread all over of the world and now it is the dominant variant in the world, which account for more than 90% of the new cases. Omicron variant spike protein carries around 30 amino acid changes, including mutations, deletions and insertions, 15 of which are in the receptor binding domain (RBD). Enhanced transmission of the Omicron variant was observed globally, which is at least 70 times more contagious than the other variants. The Omicron variant affects the effectiveness of COVID-19 vaccine and is resistant to neutralization (monoclonal antibody treatments) to a large extent.

Specifications

Gene ID (Entrez) 43740568
Species SARS-CoV-2
Accession Number QHD43416
Common Name surface glycoprotein
Concentration 200 μg/mL
Content And Storage Store SARS-CoV-2 (COVID-19) Spike G142D Δ143-145 VYY Peptide (Omicron) (at -20°C, stable for one year.)
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol S
Quantity 0.05 mg
Sequence 12 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) Spike G142D Δ143-145 VYY (Omicron).
Synonym SARS-CoV-2 Omicron Variant Spike antibody: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), Surface Glycoprotein, Spike protein, Omicron Variant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.