Learn More
Cayman Chemical SARS-CoV-2 Envelope Protein
SDP
Catalog No. 502943163
Encompass_Preferred
Source: Recombinant SARS-CoV-2 N-terminal His- and MBP-tagged E protein (C40A, C43A, C44A mutant) expressed in E. coli • Amino acids: 1-475 (full length) • MW: 52.2 kDa
Catalog No. 50-294-3163 Supplier Cayman Chemical Supplier No. 40875100 UG
Source: Recombinant SARS-CoV-2 N-terminal His- and MBP-tagged E protein (C40A, C43A, C44A mutant) expressed in E. coli • Amino acids: 1-475 (full length) • MW: 52.2 kDa