Learn More
Medchemexpress LLC SARS-CoV-2-IN-6 | 2725749-22-2 | 99.9% | 312.75 g·mol-1 | C17H13ClN2O2 | 5 MG
SDP
Catalog No. 5000262133
Encompass_Preferred
SARS-CoV-2-IN-6 is a research-use small-molecule inhibitor targeting the SARS-CoV-2 3C-like protease (3CLpro). It inhibits protease enzymatic activity with a reported IC50 of 73 nM, is supplied as a small milligram pack for laboratory studies, and is accompanied by standard product documentation and safety information.
  • Potent enzymatic inhibition with IC50 of 73 nM.
  • Designed to target the SARS-CoV-2 3C-like protease (main protease).
  • Molecular formula C17H13ClN2O2 and molecular weight 312.75 g·mol-1.
  • High purity suitable for research applications (≈99.9% by HPLC).
  • Provided in small research-scale pack sizes for assay development.
  • Intended for laboratory research use only; not for human therapeutic use.
Catalog No. 50-002-62133 Supplier Medchemexpress LLC Supplier No. HY1328865MG
SARS-CoV-2-IN-6 is a research-use small-molecule inhibitor targeting the SARS-CoV-2 3C-like protease (3CLpro). It inhibits protease enzymatic activity with a reported IC50 of 73 nM, is supplied as a small milligram pack for laboratory studies, and is accompanied by standard product documentation and safety information.
  • Potent enzymatic inhibition with IC50 of 73 nM.
  • Designed to target the SARS-CoV-2 3C-like protease (main protease).
  • Molecular formula C17H13ClN2O2 and molecular weight 312.75 g·mol-1.
  • High purity suitable for research applications (≈99.9% by HPLC).
  • Provided in small research-scale pack sizes for assay development.
  • Intended for laboratory research use only; not for human therapeutic use.