Learn More
Medchemexpress LLC SARS-CoV-2-IN-6 | 2725749-22-2 | 99.9% | C17H13ClN2O2 | 10MG
SDP
Catalog No. 5000259836
Encompass_Preferred
SARS-CoV-2-IN-6 is a small-molecule inhibitor of the SARS-CoV-2 3C-like protease with potent enzymatic activity (IC50 = 73 nM). Supplied as a high-purity solid for laboratory research, the compound is accompanied by a datasheet and safety data sheet and is intended for in vitro biochemical and antiviral mechanism studies.
  • Potent SARS-CoV-2 3CLpro inhibition (IC50 = 73 nM).
  • High purity (~99.9%) suitable for biochemical assays.
  • Solid, off-white to light yellow appearance.
  • Provided with datasheet and safety data sheet for safe handling.
  • Molecular weight 312.75 g/mol; formula C17H13ClN2O2.
  • Intended for research use only (not for human or diagnostic use).
Catalog No. 50-002-59836 Supplier Medchemexpress LLC Supplier No. HY13288610MG
SARS-CoV-2-IN-6 is a small-molecule inhibitor of the SARS-CoV-2 3C-like protease with potent enzymatic activity (IC50 = 73 nM). Supplied as a high-purity solid for laboratory research, the compound is accompanied by a datasheet and safety data sheet and is intended for in vitro biochemical and antiviral mechanism studies.
  • Potent SARS-CoV-2 3CLpro inhibition (IC50 = 73 nM).
  • High purity (~99.9%) suitable for biochemical assays.
  • Solid, off-white to light yellow appearance.
  • Provided with datasheet and safety data sheet for safe handling.
  • Molecular weight 312.75 g/mol; formula C17H13ClN2O2.
  • Intended for research use only (not for human or diagnostic use).