Learn More
Medchemexpress LLC Sars-cov-2-in-7 | 2570461-66-2 | 98.8% | C14H8F3NOSe | 50 MG
SDP
Catalog No. 5000354599
Encompass_Preferred
SARS-CoV-2-IN-7 inhibits viral replication in SARS-CoV-2-infected Vero E6 cells. It is intended for research use only.
  • Inhibits viral replication with an IC50 value of 844 nM.
  • Available as a solid, light yellow to orange in appearance.
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month.
Catalog No. 50-003-54599 Supplier Medchemexpress LLC Supplier No. HY14184150MG
SARS-CoV-2-IN-7 inhibits viral replication in SARS-CoV-2-infected Vero E6 cells. It is intended for research use only.
  • Inhibits viral replication with an IC50 value of 844 nM.
  • Available as a solid, light yellow to orange in appearance.
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month.