Learn More
Medchemexpress LLC SARS-CoV-2-IN-7 | 2570461-66-2 | 99.4% | C14H8F3NOSe | 10MG
SDP
Catalog No. 5000258931
Encompass_Preferred
SARS-CoV-2-IN-7 is a small-molecule research inhibitor that reduces SARS-CoV-2 replication in Vero E6 cells (IC50 = 844 nM). It is provided in solid and solution formats with high reported purity for laboratory research use only.
  • Reduces viral replication in cell culture (IC50 844 nM).
  • High reported purity (99.4%).
  • Available as solid samples and 10 mM DMSO solutions.
  • Multiple pack sizes from free sample to larger solids by quote.
  • Molecular formula C14H8F3NOSe; molecular weight 342.17.
  • Intended for research use only, not for clinical or therapeutic use.
Catalog No. 50-002-58931 Supplier Medchemexpress LLC Supplier No. HY14184110MG
SARS-CoV-2-IN-7 is a small-molecule research inhibitor that reduces SARS-CoV-2 replication in Vero E6 cells (IC50 = 844 nM). It is provided in solid and solution formats with high reported purity for laboratory research use only.
  • Reduces viral replication in cell culture (IC50 844 nM).
  • High reported purity (99.4%).
  • Available as solid samples and 10 mM DMSO solutions.
  • Multiple pack sizes from free sample to larger solids by quote.
  • Molecular formula C14H8F3NOSe; molecular weight 342.17.
  • Intended for research use only, not for clinical or therapeutic use.