Learn More
Medchemexpress LLC SARS-CoV-2-IN-86 (andrographolide derivative) | 368866-07-3 | MFCD02683117 | 99.4% | 466.53 g·mol⁻¹ | C26H30N2O6 | 250 MG
SDP
Catalog No. 5000302822
Encompass_Preferred
SARS-CoV-2-IN-86 is an andrographolide-derived small-molecule inhibitor of SARS-CoV-2 methyltransferases nsp14 and nsp16, characterized for in vitro activity and low predicted toxicity; supplied for research use with manufacturer guidance on solubility and storage.
  • Inhibits SARS-CoV-2 methyltransferases nsp14 and nsp16.
  • Andrographolide derivative scaffold suitable for antiviral research.
  • High purity (≈99.4%) suitable for biochemical and cellular assays.
  • Soluble in DMSO at ~50 mg/mL; may require ultrasonic and warming.
  • Stable as powder when stored at -20°C or 4°C according to guidance.
  • Available in multiple pack sizes to support different study scales.
Catalog No. 50-003-02822 Supplier Medchemexpress LLC Supplier No. 250MGHYW024485
SARS-CoV-2-IN-86 is an andrographolide-derived small-molecule inhibitor of SARS-CoV-2 methyltransferases nsp14 and nsp16, characterized for in vitro activity and low predicted toxicity; supplied for research use with manufacturer guidance on solubility and storage.
  • Inhibits SARS-CoV-2 methyltransferases nsp14 and nsp16.
  • Andrographolide derivative scaffold suitable for antiviral research.
  • High purity (≈99.4%) suitable for biochemical and cellular assays.
  • Soluble in DMSO at ~50 mg/mL; may require ultrasonic and warming.
  • Stable as powder when stored at -20°C or 4°C according to guidance.
  • Available in multiple pack sizes to support different study scales.