Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas Liat SARS-CoV-2 + Influenza A/B Control Kit

Catalog No. 23312206 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
3 Sets of Controls/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-206 3 Sets of Controls/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-206 Supplier Roche Diagnostics Supplier No. 09211128190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

The cobas SARS-CoV-2 & Influenza A/B nucleic acid test is a multiplex real-time polymerase chain reaction (PCR) test that detects and differentiates SARS-CoV-2, Influenza A, and Influenza B. This control kit is for running a control test/lot validation for the assay.

  • One sample one test, 20 minutes, know if it's COVID-19 or flu
  • Take the workload out of testing with a simple intuitive user handling
  • Minimizes cross-contamination and risk of exposure during testing with the closed-system design
  • Control Kit for use with SARS-CoV-2 & Influenza A/B Assay
  • SARS-CoV-2: 100% Positive Agreement, 100% Negative Agreement*

*Compared to an FDA-cleared EUA, cobas SARS-CoV-2 Test on cobas 6800/8800 systems

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Control Sets This control product to be used with cobas™ SARS-CoV-2 & Influenza A/B assay
CE Marker Yes
Clia Complexity Waived
Type Control
Disposable Single-use
For Use With (Application) RT-PCR
Packaging Type 1 Kit
Quantity 3 Sets of Controls/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.