Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SARS-CoV-2 (Isolate: Hong Kong/VM20001061/2020) Culture Fluid (Heat Inactivated)
SDP

Catalog No. 22158378 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-378 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-378 Supplier ZeptoMetrix Supplier No. 0810590CFHI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SARS-CoV-2 (Isolate: Hong Kong/VM20001061/2020) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.

  • Heat inactivated viral culture fluids are sold as consumable testing materials
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage
  • Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays

Specifications

Content And Storage Viral culture fluids should be stored at -65°C or below.
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Type Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Includes Each frozen aliquot contains 1 mL of titered viral culture fluid.
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.